EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and evaluation of the anti-nociceptive and anti-inflammatory activity of 4-aminoquinoline derivatives.
PMID 26116178
Abstract
In this paper, we describe the synthesis and pharmacological evaluation of a series of 4-aminoquinolines. The compounds were characterised and tested in models of pain and inflammation, using the writhing test with acetic acid, formalin test, peritonitis test by zymosan and arthritis test with Freund's adjuvant complete assay. The results revealed that all of the 4-aminoquinolines that were prepared promoted anti-nociceptive activity as well as acute and chronic anti-inflammatory effects, with marked activity in the derivates labelled with BAQ and 7-CF3-MAQ. After 7 days of treatment, 7-CF3-MAQ did not induce significant hepatotoxicity, gastrotoxicity or nephrotoxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
718335
2-Aminoquinoline, 97%
C9H8N2