EMAIL THIS PAGE TO A FRIEND
Bioorganic chemistry
Synthesis of fused tricyclic peptides using a reprogrammed translation system and chemical modification.
PMID 26117092
Abstract
Here we report a unique method of ribosomally synthesizing fused tricyclic peptides. Flexizyme-assisted in vitro translation of a linear peptide with the N-terminal chloroacetyl group and four downstream cysteines followed by the addition of 1,3,5-tris(bromomethyl)benzene results in selective production of the fused tricyclic peptide. This technology can be used for the ribosomal synthesis of fused tricyclic peptide libraries for the in vitro selection of bioactive peptides with tricyclic topology.