EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Comparative studies of pharmacokinetics and anticoagulatory effect in rats after oral administration of Frankincense and its processed products.
PMID 26117531
Abstract
Frankincense (FRA), Ruxiang, is the resin of Boswellia carterii Birdw and Boswellia bhaw-dajiana Birdw which has been used for centuries as formulas to improve the circulation and to relieve pain against carbuncles. Stir-fried Frankincense (SFF) and vinegar processed Frankincense (VPF) are two major processed Frankincense, and the processing procedures reportedly enhance the curative efficacy or reduce the side effects of FRA. This paper describes the comparisons in plasma pharmacokinetic behaviors of 11-keto-β-boswellic acid (KBA) and 3-acetyl-11-keto-β-boswellic acid (AKBA) in FRA and its processed products, and their effects on coagulation factors and blood clotting tetrachoric, using an acute cold blood-stasis animal model after oral administration of FRA, SFF, and VPF. For pharmacokinetic study, Sprague-Dawley (SD) rats were randomly divided into three groups, including group FRA, group SFF and group VPF. And the plasma samples were analyzed by HPLC. For study of anticoagulatory effect, SD rats were randomly divided into six groups, including control, acute cold blood-stasis model, Fu-fang-dan-shen tablet- (0.75g/kg), FRA-, SFF-, and VPF-treated (2.7g/kg) groups, respectively. The serum contents of thrombin-antithrombin complex (TAT), D-dimer (D-D), and prostacyclin (PGI2) of each group were measured by ELISA. The values of prothrombin time (PT), thrombin time (TT), activated partial thromboplastin time (APTT) and fibrinogen (FIB) were also assessed by hematology analyzer. Significantly increased levels of Cmax, AUC, T1/2, and MRT were found in rats treated with the processed products. In addition, decreased levels of D-D and TAT and increased contents of PGI2 were observed in rats given FRA and its processed products, compared with that of the model group. Moreover, VPF improved anticoagulation more than SFF in the animals. The observed improvement of anticoagulation by processed FRA may result from the increased absorption and bioavailability of triterpenoids.