EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Molecularly imprinted electrochemical sensor for the highly selective and sensitive determination of melamine.
PMID 26117777
Abstract
A selective and sensitive molecularly imprinted electrochemical sensor was developed for the determination of melamine. Carbon nanotube-ionic liquid composite was used as a carrier to enhance the number of imprinted cavities and then improve the selectivity and sensitivity of the sensor. The proposed sensor showed a linear relationship with the concentration of melamine in the range of 0.4 to 9.2 μM, with a detection limit of 0.11 μM. The sensor was successfully applied to the determination of melamine in milk products with good recovery.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2659
Melamine, 99%
C3H6N6