Pathology oncology research : POR
Correlation of High Mobility Group Box-1 Protein (HMGB1) with Clinicopathological Parameters in Primary Retinoblastoma.
PMID 26118980
HMGB1 is considered to be DNA chaperone as it binds without any specificity. It is the structural protein which alters nuclear homeostasis and genomic stability of chromatin. Its role in retinoblastoma (Rb) remains unclear. The aim of the present study was to evaluate the expression of HMGB1 protein in primary enucleated retinoblastomas. Expression of HMGB1 in 69 prospective cases of primary retinoblastoma were assessed by immunohistochemistry and reverse transcriptase PCR (RT-PCR) technique and correlated with clinicopathological parameters. Immunohistochemical staining revealed expression of HMGB1 in 55.07 % (38/69) cases. Semi-quantitative RT-PCR was performed on 31 fresh tumor tissues. mRNA expression was observed in 77.41 % (24/31) cases. Expression of HMGB1 was statistically significant with poor tumor differentiation (p = 0.0440) & optic nerve invasion (p = 0.0128). HMGB1 expression was frequently seen in poorly differentiated tumors and those with histopathological high risk factors. Therefore, HMGB1 may contribute to tumor invasiveness and could serve as a poor prognostic marker in Rb.