EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Protective effect of crocetin against burn-induced intestinal injury.
PMID 26119275
Abstract
Oxidative stress and inflammation exert central roles in burn-induced intestinal injury. Crocetin, a natural carotenoid compound from gardenia fruits and saffron, has been shown to inhibit oxidative stress and inflammatory response. However, the possibility of crocetin to be used in the treatment of intestinal injury after burn injury has not been investigated. The purpose of the present study was to investigate the effects and potential mechanisms of crocetin in burn-induced intestinal injury. Several free radical-generating and lipid peroxidation models were used to systematically assess the antioxidant activities of crocetin in vitro. A common burn model was used to induce the intestinal injury in rats. Changes in the levels of malondialdehyde, superoxidase dismutase, catalase, glutathione peroxidase, tumor necrosis factor α, interleukin 6, polymorphonuclear neutrophil accumulation, intestinal permeability, and intestinal histology were examined. In several models of antioxidant activity, crocetin exhibited marked inhibitory action against free radicals and lipid peroxidation. Crocetin increased levels of antioxidant enzymes and reduced intestinal oxidative injury in burn models. In addition, crocetin inhibited polymorphonuclear neutrophil accumulation, ameliorated tumor necrosis factor α and interleukin 6 levels, intestinal permeability, and histological changes. Crocetin treatment may protect against burn-induced small intestinal injury, possibly by inhibiting burn-induced oxidative stress and inflammatory response.