EMAIL THIS PAGE TO A FRIEND
Scientific reports
Investigation of multipotent postnatal stem cells from human maxillary sinus membrane.
PMID 26119339
Abstract
Maxillary sinus membrane (MSM) elevation is a common surgical technique for increasing bone height in the posterior maxilla prior to dental implant placement. However, the biological nature of bone regeneration in MSM remains largely unidentified. In this study, MSM tissue was obtained from 16 individuals during orthognathic surgery and used to isolate MSM stem cells (MSMSCs) by single-colony selection and STRO-1 cell sorting. The cell characteristics in terms of colony-forming ability, cell surface antigens, multi-differentiation potential and in vivo implantation were all evaluated. It was found that MSMSCs were of mesenchymal origin and positive for mesenchymal stem cell (MSC) markers such as STRO-1, CD146, CD29 and CD44; furthermore, under defined culture conditions, MSMSCs were able to form mineral deposits and differentiate into adipocytes and chondrocytes. When transplanted into immunocompromised rodents, MSMSCs showed the capacity to generate bone-like tissue and, importantly, maintain their MSC characteristics after in vivo implantation. These findings provide cellular and molecular evidence that MSM contains stem cells that show functional potential in bone regeneration for dental implant.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0732
Cetylpyridinium chloride, meets USP testing specifications
C21H38ClN · H2O