EMAIL THIS PAGE TO A FRIEND
Clinical biochemistry
Plasma membrane potential of lymphocytes from ataxia telangiectasia patients.
PMID 2611994
Abstract
The plasma membrane potential of lymphocytes prepared from ataxia telangiectasia (AT) patients and normal subjects was assessed using the optical indicator bis-(3-phenyl-5-oxoisoxazol-4-yl) pentamethineoxonol (oxonol-V). AT lymphocytes had a potential of -46 +/- 9 mV and normal lymphocytes had a potential of -63 +/- 4 mV. The intracellular cation content (Na+ and K+) of AT and normal lymphocytes was similar. AT and normal lymphocytes were both depolarized by extracellular K+ and to a similar extent. This study indicates that one feature characterizing ataxia telangiectasia is a modification of the ability of the lymphocyte cell membrane to sustain a normal membrane potential.