EMAIL THIS PAGE TO A FRIEND
Nature neuroscience
Different immune cells mediate mechanical pain hypersensitivity in male and female mice.
PMID 26120961
Abstract
A large and rapidly increasing body of evidence indicates that microglia-to-neuron signaling is essential for chronic pain hypersensitivity. Using multiple approaches, we found that microglia are not required for mechanical pain hypersensitivity in female mice; female mice achieved similar levels of pain hypersensitivity using adaptive immune cells, likely T lymphocytes. This sexual dimorphism suggests that male mice cannot be used as proxies for females in pain research.