EMAIL THIS PAGE TO A FRIEND
Inflammation
Cytokine Responses to Specific Immunotherapy in House Dust Mite-Induced Allergic Rhinitis Patients.
PMID 26122703
Abstract
Allergen-specific immunotherapy is the only immunomodulatory treatment that may alter the natural course of allergic disease. However, cytokine responses accompanying sublingual immunotherapy (SLIT) responder phenotypes have not been fully understood. Herein, we examined the level of crucial plasma cytokines during SLIT and evaluated whether their changes correlated to symptom scores. We observed that the levels of interleukin (IL)-17 and complement components C3a and C5a as well as IL-4 at year 3 of SLIT were significantly decreased than those at baseline. In contrast, there was no significant difference in the levels of IL-5, IL-13, and interferon (IFN)-γ. Notably, a significant positive correlation was found between the levels of IL-17 and the symptom scores at year 3. These results suggest that IL-17 could be considered a potential biomarker for the therapeutic effect of SLIT in allergic rhinitis caused by house dust mite.