EMAIL THIS PAGE TO A FRIEND
Journal of reproductive immunology
The CD200 tolerance-signaling molecule and its receptor, CD200R1, are expressed in human placental villus trophoblast and in peri-implant decidua by 5 weeks' gestation.
PMID 26123445
Abstract
CD200 expression in murine trophoblast and decidua prevents semi-allogeneic and LPS-induced abortions by binding to CD200 receptor-bearing cells to suppress NK activity, induces IDO in macrophages, and promotes the generation of regulatory T cell subsets. CD200 and its receptor CD200R1 reported in 7-9 weeks' gestation human villus trophoblasts are reduced in spontaneous abortion syncytiotrophoblasts. By specific antibody staining, we find that both CD200 and CD200R1 are expressed even earlier, by 5 weeks' gestation, by villus trophoblasts and by decidual cells. Expression of CD200 was validated using two independent antibodies. CD200-CD200R1 signaling may be required for human pregnancy success.