EMAIL THIS PAGE TO A FRIEND
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society
Cyclic mechanical stimulation rescues achilles tendon from degeneration in a bioreactor system.
PMID 26123799