EMAIL THIS PAGE TO A FRIEND
Clinical Cancer Research
Targeting Acidity in Pancreatic Adenocarcinoma: Multispectral Optoacoustic Tomography Detects pH-Low Insertion Peptide Probes In Vivo.
PMID 26124201
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7907
Sodium phosphate dibasic, BioXtra, ≥99.0%
HNa2O4P
S3264
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
HNa2O4P
S5136
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
HNa2O4P
255793
Sodium phosphate dibasic, 99.95% trace metals basis
HNa2O4P
S9763
Sodium phosphate dibasic, ACS reagent, ≥99.0%
HNa2O4P
S0876
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
HNa2O4P
71636
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
HNa2O4P
71640
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
HNa2O4P
04276
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
HNa2O4P
W239901
Sodium phosphate dibasic, 99%, FG
HNa2O4P
94046
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
HNa2O4P
RDD007
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
H2NaO4P
S8282
Sodium phosphate monobasic, BioXtra, ≥99.0%
H2NaO4P
S3139
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
H2NaO4P
S5011
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
H2NaO4P
S2554
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
H2NaO4P
229903
Sodium phosphate monobasic, 99.999% trace metals basis
H2NaO4P
S0751
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
H2NaO4P
74092
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
H2NaO4P