Anticancer research
SPINK1 Promoter Variants Are Associated with Prostate Cancer Predisposing Alterations in Benign Prostatic Hyperplasia Patients.
PMID 26124326
Several studies reported that patients with benign prostatic hyperplasia (BPH) experienced a 10% increased incidence of prostate cancer (PCa) after the first 5 years of diagnosis. We investigated the association between single nucleotide polymorphisms (SNPs) in the promoter of Serine Protease Inhibitor Kazal Type 1 (SPINK1) and the increased risk of BPH and PCa. We genotyped three SNPs in a cases-control study, including BPH and PCa cases. Multiple logistic regression models were applied to analyze clinical and genotypic data. We found an inverse association between SNP rs10035432 and BPH under the log-additive (p=0.007) model. No association was found between these SNPs and PCa risk. However, we observed a possible association between rs1432982 and lower-grade PCa (p=0.05) under the recessive model. SPINK1 promoter variants are likely to be associated with the risk of BPH.