EMAIL THIS PAGE TO A FRIEND
Prenatal diagnosis
The hippocampal commissure: a new finding at prenatal 3D ultrasound in fetuses with isolated complete agenesis of the corpus callosum.
PMID 26126648
Abstract
The aim of this research was to determine the prevalence and sonographic appearance of the hippocampal commissure in fetuses with isolated complete agenesis of the corpus callosum by three-dimensional neurosonography in the multiplanar mode. This was a multicenter observational study. Stored volume datasets of fetuses with isolated complete agenesis of the corpus were retrospectively retrieved for analysis in three tertiary centers. The presence or absence of the hippocampal commissure was independently evaluated in the coronal and midsagittal planes by two operators. Postnatal follow-up was obtained in all cases. From November 2007 to February 2013, 41 cases between 19 and 30 weeks of gestation were retrieved for analysis. The hippocampal commissure was visible in the coronal and sagittal planes in 27/41 (65.8%), absent or not clearly recognizable in the remaining 14 cases. The qualitative analysis of the two operators was concordant in 100% of cases. In more than half of fetuses with complete callosal agenesis, the hippocampal commissure may be visualized at prenatal ultrasound. This is a residual interhemispheric connection, which in normal cases is hidden by the corpus callosum itself. Further research is needed to establish if this has an impact on postnatal outcome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0138
Tetrazolium Violet
C23H17ClN4