EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Detection of adenine synthesis intermediate products in the air].
PMID 2612950
Abstract
Photometric procedures of monitoring the content of adenine synthesis intermediate products were developed and optimum conditions of air sample analysis and selection determined. Thus it became possible to establish general exposure levels for (phenylazo) malondinitryl and 4,5,6 triaminopyrimidine sulfate, sensitivity of determination being 0.25 mg/l.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
307181
4,5,6-Triaminopyrimidine sulfate, 98%
C4H9N5O4S