EMAIL THIS PAGE TO A FRIEND
Journal of diabetes and its complications
Proteomic analysis of plasma proteins in diabetic retinopathy patients by two dimensional electrophoresis and MALDI-Tof-MS.
PMID 26129748
Abstract
Diabetic retinopathy is a highly specific vascular complication of diabetes mellitus and progresses from mild non-proliferative abnormalities characterized by increased vascular permeability to moderate and severe proliferative diabetic retinopathy characterized by the growth of blood vessels on the retina. The aim of the study was to identify the differentially expressed proteins in diabetic retinopathy using two-dimensional electrophoresis. Blood sample was drawn from subjects with diabetes mellitus (without retinopathy) who served as controls and patients with diabetic retinopathy in tubes containing EDTA as anticoagulant. Albumin and immunoglobulin IgG collectively removed to enrich proteins of lower abundance. 2de was carried out to see if there are any differentially expressed proteins. Approximately 48 and 61 spots were identified in control and diabetic retinopathy respectively, of which three protein spots RBP1 (retinol-binding protein 1), NUD10 (Diphosphoinositol polyphosphohydrolase 3 alpha), NGB (neuroglobin) were down regulated and HBG2 (hemoglobin) and BY55 (CD 160 antigen) were upregulated in diabetic retinopathy. These five protein spots were excised and were subjected to in-gel tryptic digestion, and their identities were determined by ultraflex MALDI-TOF-MS. We report a comprehensive patient-based plasma proteomic approach to the identification of potential biomarkers for diabetic retinopathy screening and detection. We identified 5 different proteins that were differentially expressed in the plasma of control diabetic patients (without retinopathy). Among these five proteins the expression of neuroglobin (NGB) protein varied significantly and may be a potential biomarker in diabetic retinopathy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
476870
α-Cyano-4-hydroxycinnamic acid, 99%
C10H7NO3