EMAIL THIS PAGE TO A FRIEND
Veterinary microbiology
Sub-inhibitory concentrations of penicillin G induce biofilm formation by field isolates of Actinobacillus pleuropneumoniae.
PMID 26130517
Abstract
Actinobacillus pleuropneumoniae is a Gram-negative bacterium and causative agent of porcine pleuropneumonia. This is a highly contagious disease that causes important economic losses to the swine industry worldwide. Penicillins are extensively used in swine production and these antibiotics are associated with high systemic clearance and low oral bioavailability. This may expose A. pleuropneumoniae to sub-inhibitory concentrations of penicillin G when the antibiotic is administered orally. Our goal was to evaluate the effect of sub-minimum inhibitory concentration (MIC) of penicillin G on the biofilm formation of A. pleuropneumoniae. Biofilm production of 13 field isolates from serotypes 1, 5a, 7 and 15 was tested in the presence of sub-MIC of penicillin G using a polystyrene microtiter plate assay. Using microscopy techniques and enzymatic digestion, biofilm architecture and composition were also characterized after exposure to sub-MIC of penicillin G. Sub-MIC of penicillin G significantly induced biofilm formation of nine isolates. The penicillin G-induced biofilms contained more poly-N-acetyl-D-glucosamine (PGA), extracellular DNA and proteins when compared to control biofilms grown without penicillin G. Additionally, penicillin G-induced biofilms were sensitive to DNase which was not observed with the untreated controls. Furthermore, sub-MIC of penicillin G up-regulated the expression of pgaA, which encodes a protein involved in PGA synthesis, and the genes encoding the envelope-stress sensing two-component regulatory system CpxRA. In conclusion, sub-MICs of penicillin G significantly induce biofilm formation and this is likely the result of a cell envelope stress sensed by the CpxRA system resulting in an increased production of PGA and other matrix components.