EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental medicine
Peptic ulcer hemorrhage combined with acute gout: analyses of treatment in 136 cases.
PMID 26131224
Abstract
This study aims to compare the safety and curative effect of celecoxib and small-dose methylprednisolone sodium succinate in patients with peptic ulcer hemorrhage combined with acute gout. In this randomized, controlled trial, a total of 136 patients with peptic ulcer hemorrhage combined with acute gout were divided into the celecoxib group or the small-dose methylprednisolone sodium succinate group. These patients underwent gastroscopy hemostasis and proton pump inhibitor (PPI) therapy. Moreover, for the treatment of gout, the patients were administered either celecoxib or small-dose methylprednisolone sodium succinate. Adverse reactions and the visual analogue scale (VAS) score were recorded for the two groups. The difference in adverse reactions between the two groups was not significant (χ(2) = 0.002, P = 0.967). The duration of evident pain relief after the first dose of treatment showed a significant difference between the two groups (t = 13.728, P < 0.01). The VAS scores before treatment were not significantly different between the two groups (t = -1.786, P = 0.076). The VAS scores at 6 h, 2 days, 4 days, 6 days, and 8 days after treatment were significantly different between the two groups (t = 3.239, 6.586, 6.280, 3.737, 3.215; P = 0.002, 0.000, 0.000, 0.000, 0.002, respectively). In cases that receive effective gastroscopy hemostasis and PPI therapy, small-dose methylprednisolone sodium succinate exhibits a greater clinical curative effect for peptic ulcer hemorrhage combined with acute gout as compared to celecoxib, and is associated with greater safety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W327700
Disodium succinate, ≥98%
C4H4Na2O4