EMAIL THIS PAGE TO A FRIEND
Basic & clinical pharmacology & toxicology
Potentiation of Glibenclamide Hypoglycaemia in Mice by MK-467, a Peripherally Acting Alpha2-Adrenoceptor Antagonist.
PMID 26132275
Abstract
Pharmacological antagonism and genetic depletion of pancreatic α2A-adrenoceptors increase insulin secretion in mice and enhance the insulinotropic action of glibenclamide, a representative of the sulphonylurea class of insulin secretagogues used in the therapy of type 2 diabetes. Antagonism of α2-adrenoceptors in the central nervous system (CNS) causes tachycardia and hypertension, making generalized α2-adrenoceptor blockade unfavourable for clinical use despite its potential to decrease blood glucose levels. The purpose of this study was to test the acute effects of the peripherally acting α2-adrenoceptor antagonist MK-467 alone and in combination with glibenclamide in non-diabetic C57BL/6N mice. Cardiovascular safety was assessed in freely moving mice with radiotelemetry. Dose-dependent decreases in blood glucose and increases in plasma insulin concentrations were seen with the combination of MK-467 and glibenclamide; the combinations were much more potent than glibenclamide or MK-467 alone. Furthermore, MK-467 had no effect on mean arterial pressure or heart rate in freely moving mice and did not prevent the centrally mediated hypotensive effect of the α2-adrenoceptor agonist medetomidine. Thus, peripheral blockade of α2-adrenoceptors does not evoke the same cardiovascular adverse effects as antagonism of CNS α2-adrenoceptors. The current results indicate that the combined use of small doses of a peripherally acting α2-adrenoceptor antagonist with a sulphonylurea drug could provide a novel option for the treatment of type 2 diabetes, especially in patients with increased tonic α2-adrenoceptor-mediated inhibition of insulin secretion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9611
Atipamezole, ≥98% (HPLC)
C14H16N2