EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
The Effect of Diagnostic Absorbed Doses from 131I on Human Thyrocytes in Vitro.
PMID 26132566
Abstract
Administration of diagnostic activities of 131I, performed in order to detect thyroid remnants after surgery and/or thyroid cancer recurrence/metastases, may lead to reduction of iodine uptake. This phenomenon is called "thyroid stunning". We estimated radiation absorbed dose-dependent changes in genetic material, in particular in sodium iodide symporter (NIS) gene promoter, and NIS protein level in human thyrocytes (HT). We used unmodified HT isolated from patients subjected to thyroidectomy exposed to 131I in culture. The different 131I activities applied were calculated to result in absorbed doses of 5, 10, and 20 Gy. According to flow cytometry analysis and comet assay, 131I did not influence the HT viability in culture. Temporary increase of 8-oxo-dG concentration in HT directly after 24 h (p < 0.05) and increase in the number of AP-sites 72 h after termination of exposition to 20 Gy dose (p < 0.0001) were observed. The signs of dose-dependent DNA damage were not associated with essential changes in the NIS expression on mRNA and protein levels. Our observation constitutes a first attempt to evaluate the effect of the absorbed dose of 131I on HT. The results have not confirmed the theory that the "thyroid stunning" reduces the NIS protein synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220051
N-Iodosuccinimide, 95%
C4H4INO2