EMAIL THIS PAGE TO A FRIEND
Oncology reports
Suppression of KLF8 induces cell differentiation and sensitizes colorectal cancer to 5-fluorouracil.
PMID 26133391
Abstract
KLF8 is a member of the KLF transcription factor family that plays an important role in oncogenesis. However, the role of KLF8 in colorectal cancer remains unknown. The aims of the present study were to examine KLF8 expression in colorectal cancers, to determine the role of KLF8 in cell differentiation and to investigate the antiproliferative effect of KLF8 silencing. The expression of KLF8 and phospho-ERK proteins was analyzed, and the effects of KLF8 suppression on cell differentiation and growth were evaluated. In addition, the biological impact of KLF8 knockdown on colorectal cancer cells was investigated inxa0vitro and inxa0vivo. The expression of the KLF8 protein was higher in 10/14 (71.43%) fresh cancer tissues compared with adjacent normal tissues, and the blockade of ERK signaling by U0126 decreased the expression of KLF8 in a time-xa0and dose-dependent manner. Furthermore, KLF8-siRNA induced the expression of carcinoembryonic antigen (CEA) and E-cadherin as well as the maturation of F-actin. KLF8 suppression inhibited serum-dependent, anchorage‑dependent and -independent cell growth. Moreover, KLF8 silencing induced apoptosis and sensitized cancer cells to 5-fluorouracil (5-FU). A strong antitumorigenic effect by lenti-KLF8-shRNA, which was enhanced when combined with 5-FU treatment, was exerted in nude mice. Thus, KLF8 suppression induced cell differentiation and inhibited tumorigenesis.