EMAIL THIS PAGE TO A FRIEND
Journal of oleo science
Competitive Solubilization of Cholesterol/Cholesteryl Oleate and Seven Species of Sterol/Stanol in Model Intestinal Solution System.
PMID 26136176
Abstract
The addition of plant sterols/stanols (sterols or stanols) can reduce the solubilization of cholesterol in a model intestinal solution system. We studied the molecular structure of seven different sterols/stanols and the effect they had on the solubilization of cholesterol or cholesterol ester in a model intestinal solution. The differences in the molecular structures of the sterol/stanol species influenced their abilities to reduce the solubility of cholesterol in the competitive solubilization experiments. Cholestanol whose molecular structure resembled cholesterol was the most effective at reducing the solubilization of cholesterol and cholesterol ester, with the solubilities of cholesterol and cholesteryl oleate being 41% and 39% respectively of the values observed for the single solubilizate systems. β-Sitosterol was also able to reduce the solubilities of cholesterol and cholesteryl oleate to 43% and 45% of those observed in a single solubilizate system. Both, stigmasterol and brassicasterol have an unsaturated double bond in a steroid side chain and did not exhibit major cholesterol-lowering effects. These results were reflected by the Gibbs free energy change values (ΔG(0)) for solubilization, where the sterol/stanol species with cholesterol-lowering effects had similar or larger negative ΔG(0) values than those observed for cholesterol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9253
Cholesteryl oleate, ≥98% (HPLC; detection at 205 nm)
C45H78O2
T6260
Sodium taurochenodeoxycholate
C26H44NNaO6S