EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Impairment of the Cellular Distribution and Stability of the Erythropoietin Receptor Through the Direct Targeting of Aristolochic Acid.
PMID 26136230
Abstract
Aristolochic acid (AA) nephropathy is complicated with early onset and severe anemia. The molecular pathological mechanism of AA-induced anemia remains unclear. The aim of this study was to evaluate the putative pathological roles of the erythropoietin receptor (EPOR) in AA-induced anemia in both AA nephropathy zebrafish and cultured human renal tubular cells (HK2). Immunofluorescence staining experiments revealed that AA colocalizes with the EPOR in zebrafish embryos as well as in the cytoplasm of HK2 cells. After exogenous EPO stimulation, the EPOR was detected in the plasma membrane of HK cells. However, cotreatment with AA and EPO inhibited EPOR signaling and its membrane localization upon EPO stimulation. The results of studies with a protein synthesis inhibitor and a lysosome inhibitor revealed that AA accelerates the lysosomal degradation of EPOR. The molecular docking results suggest that AA may interact with the N-terminus of EPOR. Together with the results of light absorption and inxa0vitro competition assays, we concluded that AA treatment impairs EPOR membrane localization, accelerates its lysosomal degradation, and consequently downregulates EPOR signaling by direct targeting. The results of this study may further detail the pathological mechanism of severe anemia complicated with AA nephropathy.