EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Inhibiting an Epoxide Hydrolase Virulence Factor from Pseudomonas aeruginosa Protects CFTR.
PMID 26136396
Abstract
Opportunistic pathogens exploit diverse strategies to sabotage host defenses. Pseudomonas aeruginosa secretes the CFTR inhibitory factor Cif and thus triggers loss of CFTR, an ion channel required for airway mucociliary defense. However, the mechanism of action of Cif has remained unclear. It catalyzes epoxide hydrolysis, but there is no known role for natural epoxides in CFTR regulation. It was demonstrated that the hydrolase activity of Cif is strictly required for its effects on CFTR. A small-molecule inhibitor that protects this key component of the mucociliary defense system was also uncovered. These results provide a basis for targeting the distinctive virulence chemistry of Cif and suggest an unanticipated role of physiological epoxides in intracellular protein trafficking.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5752
Adrenochrome
C9H9NO3
308323
cis-Stilbene oxide, 97%
C14H12O
E1012
Epibromohydrin, 98%
C3H5BrO