EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
CD4
PMID 26137395
Abstract
Chronic pancreatitis (CP) is a risk factor of pancreatic ductal adenocarcinoma (PDAC) and characterized by a pronounced desmoplastic reaction with CD4