EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
Metronomic cyclophosphamide eradicates large implanted GL261 gliomas by activating antitumor Cd8
PMID 26137402
Abstract
Cancer chemotherapy using cytotoxic drugs can induce immunogenic tumor cell death; however, dosing regimens and schedules that enable single-agent chemotherapy to induce adaptive immune-dependent ablation of large, established tumors with activation of long-term immune memory have not been identified. Here, we investigate this issue in a syngeneic, implanted GL261 glioma model in immune-competent mice given cyclophosphamide on a 6-day repeating metronomic schedule. Two cycles of metronomic cyclophosphamide treatment induced sustained upregulation of tumor-associated CD8
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8031
N6-Cyclopentyladenosine, solid
C15H21N5O4