EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
CD20
PMID 26137418
Abstract
Recently, a small subset of T cells that expresses the B cell marker CD20 has been identified in healthy volunteers and in patients with rheumatoid arthritis and multiple sclerosis. The origin of these CD20-positive T cells as well as their relevance in human disease remains unclear. Here, we identified that after functional B cell/T cell interaction CD20 molecules are transferred to the cell surface of T cells by trogocytosis together with the established trogocytosis marker HLA-DR. Further, the presence of CD20 on isolated CD20