EMAIL THIS PAGE TO A FRIEND
Nanoscale
Layered titanium diboride: towards exfoliation and electrochemical applications.
PMID 26137856
Abstract
Layered transition metal diborides (TMDB), amongst other refractory metal borides, are commonly employed for material fabrication such as wear- and corrosion-resistant coatings due to their impressive chemical stability and thermal conductivity. In spite of the wide scope of studies carried out on TMDB in the physical field, investigations on its electrochemistry remain limited. Since the physical properties play a vital role in any material's electrochemical behaviour, we explore the viability of the most popular form of titanium boride, layered TiB2, as catalysts for electrochemical energy reactions, including hydrogen evolution and oxygen reduction. Three types of TiB2 were compared in this work - TiB2 separately modified with sodium naphtalenide and butyllithium in an attempt to exfoliate TiB2 and unmodified TiB2. The electrocatalytic activity displayed by all three TiB2 materials provides a wider range of opportunities for the application of TiB2 in material studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
427551
Benzophenone, purified by sublimation, ≥99%
C13H10O
239852
Benzophenone, ReagentPlus®, ≥99%
C13H10O
B9300
Benzophenone, ReagentPlus®, 99%
C13H10O
W213403
Benzophenone, ≥99%, FCC, FG
C13H10O
B3634
Benzophenone
C13H10O
GF93740384
Carbon - Vitreous, rod, 100mm, diameter 10mm, glassy carbon
C
GF65427587
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
C
GF05962765
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
C
GF68195403
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
C
GF23723423
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
C
GF91785134
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
C
GF78447707
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
C
GF45345562
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
C
GF24425805
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
C
GF93251011
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
C
GF20176680
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
C
GF59400233
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF39088648
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF07378476
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF39873369
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF61342220
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF58042324
Carbon - Vitreous, foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF12408713
Carbon - Vitreous, foam, 275x330mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF04031132
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF06230922
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF95357526
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
C
GF61808889
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
C
GF09325009
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
C
GF22691426
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
C
GF74135628
Carbon - Vitreous, foil, 10x10mm, thickness 2.0mm, glassy carbon
C