EMAIL THIS PAGE TO A FRIEND
Environmental Toxicology and Chemistry / Setac
Sublethal health effects in laboratory rodents from environmentally relevant exposures to oil sands contaminants.
PMID 26139097