EMAIL THIS PAGE TO A FRIEND
Environmental Science and Pollution Research International
PAH occurrence in chalk river systems from the Jura region (France). Pertinence of suspended particulate matter and sediment as matrices for river quality monitoring.
PMID 26139398
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48567
Anthracene, analytical standard
C14H10
40076
Anthracene solution, certified reference material, 1000 μg/mL in acetone
C14H10
48647
Anthracene solution, certified reference material, 200 μg/mL in methanol
C14H10
B1760
Benzo[a]pyrene, ≥96% (HPLC)
C20H12
48564
Benzo[a]pyrene, analytical standard, for environmental analysis
C20H12
49473-U
Benzo[a]pyrene solution, certified reference material, 100 μg/mL in methylene chloride
C20H12
CRM40071
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
C20H12
CRM48665
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
C20H12
392251
Benzo[k]fluoranthene, for fluorescence, ≥99%
C20H12
48492
Benzo[k]fluoranthene, analytical standard, This product has been replaced by product 03323-10MG
C20H12
48668
Benzo[k]fluoranthene solution, certified reference material, 200 μg/mL in methylene chloride
C20H12
40073
Benzo[k]fluoranthene solution, certified reference material, 1000 μg/mL in acetone
C20H12
245186
Chrysene, 98%
C18H12
48565-U
Chrysene, analytical standard, This product has been replaced by product 35754-100MG
C18H12
SCD-003
Dibenz[a,h]anthracene, vial of 1 g, analytical standard
C22H14
48574
Dibenz[a,h]anthracene, analytical standard
C22H14
423947
Fluoranthene, 98%
C16H10
48535
Fluoranthene, analytical standard
C16H10
F807
Fluoranthene, 98%
C16H10
ERI-001
Indeno[1,2,3-cd]pyrene, vial of 25 mg, analytical standard
C22H12
SCI-001
Indeno[1,2,3-cd]pyrene, vial of 1 g, analytical standard
C22H12
48499
Indeno[1,2,3-cd]pyrene, analytical standard
C22H12
P5891
L-Pyroglutamic acid 2-naphthylamide, ≥99% (TLC)
C15H14N2O2
394475
Perylene, sublimed grade, ≥99.5%
C20H12
P11204
Perylene, ≥99%
C20H12