EMAIL THIS PAGE TO A FRIEND
Journal of food science and technology
The physicochemical properties of legume protein isolates and their ability to stabilize oil-in-water emulsions with and without genipin.
PMID 26139878
Abstract
The physicochemical and emulsifying properties of legume protein isolates prepared from chickpea (CPI), faba bean (FPI), lentil (LPI) and soy (SPI) were investigated in the presence and absence of genipin. Solubility was highest for CPI (~94xa0%), followed by LPI (~90xa0%), FPI (~85xa0%) and SPI (~50xa0%). Surface characteristics revealed similar zeta potentials (~ - 47xa0mV) for CPI, LPI and FPI, but lower for SPI (~ - 44xa0mV). Contrastingly, surface hydrophobicity was greatest for CPI (~137 arbitrary units, AU), followed by SPI/LPI (~70xa0AU) and FPI (~24xa0AU). A significant (from 16.73 to ~8.42 mN/m) reduction in interfacial tension was observed in canola oil-water mixtures in the presence of non-crosslinked legume protein isolates. The extent of legume protein isolate-genipin crosslinking was found to be similar for all isolates. Overall, creaming stability increased in the presence of genipin, with maximum stability observed for SPI (65xa0%), followed by FPI (61xa0%), LPI (56xa0%) and finally CPI (50xa0%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1028
8-Anilino-1-naphthalenesulfonic acid
C16H13NO3S