EMAIL THIS PAGE TO A FRIEND
Journal of food science and technology
Suitable coating material for microencapsulation of spray-dried fish oil.
PMID 26139910
Abstract
This study was conducted to screen the most suitable coating material for the production of microencapsulated fish oil powder using ternary blends of maltodextrin (15, 25 % w/w), Arabic gum (2.5, 7.5 % w/w), and methylcellulose (0.5, 1.5 % w/w). The physical properties of fish oil emulsion and encapsulated powders were evaluated. Arabic gum (5 % w/w) showed the most significant (p < 0.05) effect on the surface mean diameter of the droplets in the emulsion. Maltodextrin had the most significant (p < 0.05) effect on the centrifuge stability of the emulsion and the amount of surface oil of the powder at 15 and 20 % (w/w) respectively, whereas methylcellulose (0.5 % w/w) had the most significant (p < 0.05) effect on the width distribution of the droplets in the emulsion. The total optimal area leading to the formation of coating material with desirable physical properties was expected to be obtained by the combination of 16 % (w/w) maltodextrin, 6.5 % (w/w) Arabic gum, and 0.88 % (w/w) methylcellulose respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20021
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
C11H16O2