EMAIL THIS PAGE TO A FRIEND
Avicenna journal of medical biotechnology
Comparison between Platinum-Azidothymidine and Azidothymidine Effects on Bcl-2 and Telomerase Gene Expression in Rats with Hepatocellular Carcinoma.
PMID 26140181
Abstract
High expression of telomerase and Bcl-2 are reported in hepatocellular carcinoma. Some anticancer drugs show their effects through reduction of these factors. In this study, it was aimed to investigate the effects of a new synthetic compound, platinum azidothymidine, on inhibition of telomerase and Bcl-2 expression in hepatocellular carcinoma compared to azidothymidine. To study the effects of Pt-AZT on hepatocellular carcinoma and compare its effects with AZT in inhibition of telomerase and Bcl-2 gene expression, pathogen-free male Wistar rats (n=100) were used. They were randomly divided to 4 groups (n=25). Group A as the control group contained 25 healthy rats; in the rest of animals, preneoplastic lesions were induced in their livers (groups B, C, and D) using Solt-Farber resistant hepatocyte protocol. Cancer development was approved by a pathology laboratory. Group B was negative control (untreated), groups C and D were treated by intraperitoneal injection (IP) of Pt-AZT (0.9 mg/kg/day) and AZT (0.3 mg/kg/day), respectively for 14 days. At the end of the protocol, all rats were sacrificed and Bcl-2 and telomerase gene expression was determined using real-time PCR. No tumor in the livers was found in group A at any point of the study, but it was present in livers of all animals in B, C and D groups. Results showed that telomerase and Bcl-2 expression was significantly lower in group C compared with group B (0.473±0.231 vs. 5.137±5.08, p<0.001, for telomerase expression, and 0.41±0.276 vs. 7.25±11.6, p<0.001, for Bcl-2 expression) and also compared with group D (0.473±0.231 vs. 3.48±4.02, p<0.001, for telomerase expression, and 0.41±0.276 vs. 4.93±18, p<0.001, for Bcl-2 expression). For the first time, it was demonstrated that Pt-AZT has more inhibitory effect on telomerase and Bcl-2 expression than AZT. It effectively inhibits the growth of liver tumor in rats by extending apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2169
3′-Azido-3′-deoxythymidine, ≥98% (HPLC)
C10H13N5O4