EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Integrated Transcriptional and Proteomic Analysis of Growth Hormone Suppression Mediated by Trichothecene T-2 Toxin in Rat GH3 Cells.
PMID 26141394
Abstract
Chronic exposure to trichothecenes is known to disturb insulin-like growth factor 1 and signaling of insulin and leptin hormones and causes considerable growth retardation in animals. However, limited information was available on mechanisms underlying trichothecene-induced growth retardation. In this study, we employed an integrated transcriptomics, proteomics, and RNA interference (RNAi) approach to study the molecular mechanisms underlying trichothecene cytotoxicity in rat pituitary adenoma GH3 cells. Our results showed that trichothecenes suppressed the synthesis of growth hormone 1 (Gh1) and inhibited the eukaryotic transcription and translation initiation by suppressing aminoacyl-tRNA synthetases transcription, inducing eukaryotic translation initiation factor 2-alpha kinase 2 (EIF2AK2) and reducing eukaryotic translation initiation factor 5 a. The sulfhydryl oxidases , protein disulfide isomerase,and heat shock protein 90 (were greatly reduced, which resulted in adverse regulation of protein processing and folding. Differential genes and proteins associated with a decline in energy metabolism and cell cycle arrest were also found in our study. However, use of RNAi to interfere with hemopoietic cell kinase (Hck) and EIF2AK2 transcriptions or use of chemical inhibitors of MAPK, p38, Ras, and JNK partially reversed the reduction of Gh1 levels induced by trichothecenes. It indicated that the activation of MAPKs, Hck, and EIF2AK2 were important for trichothecene-induced growth hormone suppression. Considering the potential hazards of exposure to trichothecenes, our findings could help to improve our understanding regarding human and animal health implications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8285
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
C21H27N7O14P2
N8410
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
C21H27N7O14P2
N8535
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
C21H27N7O14P2
N7004
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
C21H27N7O14P2 · xH2O
N6522
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
C21H27N7O14P2 · xH2O
N1636
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
C21H27N7O14P2 · xH2O
N3014
β-Nicotinamide adenine dinucleotide hydrate, cell culture tested, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
C21H27N7O14P2 · xH2O
N7381
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1, ≥95% (HPLC)
C21H27N7O14P2 · xH2O
N1511
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
C21H27N7O14P2 · xH2O
43410
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
C21H27N7O14P2 · xH2O
N7132
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
C21H26LiN7O14P2
D0156
Deoxynivalenol
C15H20O6
43815
DL-Dithiothreitol, BioUltra, for molecular biology, ≥99.5% (RT)
C4H10O2S2
43819
DL-Dithiothreitol, ≥99.0% (RT)
C4H10O2S2
D0632
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
C4H10O2S2
D5545
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
C4H10O2S2
D9779
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
C4H10O2S2
D9163
DL-Dithiothreitol, for electrophoresis, ≥99%
C4H10O2S2
I1149
Iodoacetamide, BioUltra
C2H4INO
I6125
Iodoacetamide, ≥99% (NMR), crystalline
C2H4INO
A3221
Iodoacetamide, Single use vial of 56 mg
C2H4INO
I5161
Iodoacetamide, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production
C2H4INO
K1385
KN-93, ≥98% (HPLC)
C26H29ClN2O4S · H3PO4
30-4310
Trifluoroacetic acid, SAJ special grade, ≥99.0%
C2HF3O2
91709
Trifluoroacetic acid solution, BioReagent, suitable for protein sequencing, ~25% in H2O
C2HO2F3