EMAIL THIS PAGE TO A FRIEND
Anaerobe
A prospective study of community-associated Clostridium difficile infection in Kuwait: Epidemiology and ribotypes.
PMID 26144314
Abstract
Clostridium difficile infection (CDI) is increasingly recognized as a significant community acquired pathogen that causes disease in the community. The aim of the study was to investigate prospectively the incidence of community-acquired-CDI (CA-CDI) in Kuwait. Of the 2584 patients with diarrhea, 16 (0.62%) were confirmed cases of CA-CDI. The other notable pathogens were Salmonella spp. (0.39%) and Campylobacter spp. (0.23%). The mean age was 39 years and the CDI was mild. Exposure to antibiotics in the previous 12 weeks, contact with infant aged <2 years and history of foreign travel was significantly associated with CA-CDI (Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6880
D-Cycloserine, powder
C3H6N2O2
C3909
D-Cycloserine, synthetic
C3H6N2O2
30020
D-Cycloserine, ≥96.0% (NT)
C3H6N2O2