EMAIL THIS PAGE TO A FRIEND
BioMed research international
Activation of Cell Surface Bound 20S Proteasome Inhibits Vascular Cell Growth and Arteriogenesis.
PMID 26146628
Abstract
Arteriogenesis is an inflammatory process associated with rapid cellular changes involving vascular resident endothelial progenitor cells (VR-EPCs). Extracellular cell surface bound 20S proteasome has been implicated to play an important role in inflammatory processes. In our search for antigens initially regulated during collateral growth mAb CTA 157-2 was generated against membrane fractions of growing collateral vessels. CTA 157-2 stained endothelium of growing collateral vessels and the cell surface of VR-EPCs. CTA 157-2 bound a protein complex (760 kDa) that was identified as 26 kDa α7 and 21 kDa β3 subunit of 20S proteasome in mass spectrometry. Furthermore we demonstrated specific staining of 20S proteasome after immunoprecipitation of VR-EPC membrane extract with CTA 157-2 sepharose beads. Functionally, CTA 157-2 enhanced concentration dependently AMC (7-amino-4-methylcoumarin) cleavage from LLVY (N-Succinyl-Leu-Leu-Val-Tyr) by recombinant 20S proteasome as well as proteasomal activity in VR-EPC extracts. Proliferation of VR-EPCs (BrdU incorporation) was reduced by CTA 157-2. Infusion of the antibody into the collateral circulation reduced number of collateral arteries, collateral proliferation, and collateral conductance in vivo. In conclusion our results indicate that extracellular cell surface bound 20S proteasome influences VR-EPC function in vitro and collateral growth in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
474711
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Ba
441880
Barium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Ba
403334
Barium, pieces, 1 cm, 99% trace metals basis
Ba
237094
Barium, rod, diam. ~2 cm, ≥99% trace metals basis
Ba
735787
Barium, beads, 0.5-2.0 mm particle size, 99% trace metals basis
Ba
B4429
BIS-TRIS, BioPerformance Certified, cell culture tested, suitable for insect cell culture, ≥98.0%
C8H19NO5
B9754
BIS-TRIS, ≥98.0% (titration)
C8H19NO5
RES1161B-A7
BIS-TRIS, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
C8H19NO5
PHG0004
BIS-TRIS, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H19NO5
B7535
BIS-TRIS, BioXtra, ≥98.0% (titration)
C8H19NO5
774472
Ethyl 2-(4-methoxyphenylcarbonothioylthio)acetate, 99%
C12H14O3S2
I1149
Iodoacetamide, BioUltra
C2H4INO
I6125
Iodoacetamide, ≥99% (NMR), crystalline
C2H4INO
A3221
Iodoacetamide, Single use vial of 56 mg
C2H4INO
I5161
Iodoacetamide, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production
C2H4INO
X1126
Xylazine, ≥99%
C12H16N2S