EMAIL THIS PAGE TO A FRIEND
Investigative Radiology
Persistent Pure Ground-Glass Nodules Larger Than 5 mm: Differentiation of Invasive Pulmonary Adenocarcinomas From Preinvasive Lesions or Minimally Invasive Adenocarcinomas Using Texture Analysis.
PMID 26146871
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5585
5-(N-Methyl-N-isobutyl)­amiloride, ≥98% (TLC), powder
C11H18ClN7O