EMAIL THIS PAGE TO A FRIEND
Medical and veterinary entomology
Life history of the sand fly vector Lutzomyia cruciata in laboratory conditions.
PMID 26147368
Abstract
Lutzomyia cruciata Coquillet (Diptera: Psychodidae: Phlebotominae) is a potential vector of Leishmania sp.; its geographical distribution in Mexico is widespread, but its life history is unknown. The present study gives relevant information on the life cycle, morphology, survival and reproduction of Lu. cruciata observed over successive generations under laboratory conditions. Seven successive generations were produced. A total of 975 adults were obtained in a sexual proportion of 1.1 : 1 (female : male). Each Lu. cruciata female produced 20.7 eggs and 1.9 adults, approximately, with a proportion of eggs per female of 2.7% (first generation) and 21.3% (second generation). The life cycle of Lu. cruciata, from egg to adult, occurred in 52.7 ± 0.52 days. The largest percentage of mortality occurred during the egg stage (48.5%) and the first larval instar (26.5%), whereas in the pupal stage mortality was the lowest (9.1%). Lutzomyia cruciata exhibits sexual dimorphism based on size, which is exhibited as of the second larval instar, males being smaller than females. The maximum survival of females and males was 10 and 15 days, respectively. An overview of the immature stages of the species made with an electronic scanning microscope is included. This paper contributes basic information on aspects of Lu. cruciata that were previously unknown related to its life history.