EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Model-based description of peptide retention on doped reversed-phase media.
PMID 26150253
Abstract
Reversed-phase (RP) chromatography is one of the main tools for the preparative purification of therapeutic peptides. In previous works [1,2], a new type of RP chromatography, doped reversed-phase chromatography (DRP) was presented. By adding small amounts (up to 15% of the surface ligands) of repulsive ion exchange ligands to a traditional RP material, significant improvements in peptide purification performance were observed, at the same or in similar operating conditions. These improvements included increases in selectivity in diluted conditions (up to twice as high), increases in yield in preparative conditions (up to 20% higher) and in productivity in preparative conditions (up to twice as high), when compared to RP materials [2]. A proper physical model is developed in this work to quantitatively explain and rationalize this behavior. The developed model is then used to correctly fit the retention data of several peptides in different buffering conditions. The increase in selectivity is related to a controlled decrease in free surface area available for adsorption due to the ionic ligands creating a repulsive sphere the analytes cannot enter. This decrease in adsorption surface is calculated using Debye-Hückel theory, and in combination with linear solvent strength theory, allows for the quantitative description of peptide retention on DRP media.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5811
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
H3O4P
466123
Phosphoric acid, crystalline, ≥99.999% trace metals basis
H3O4P
452289
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
H3O4P
345245
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
H3O4P
79617
Phosphoric acid, BioUltra, ≥85% (T)
H3O4P
40278
Phosphoric acid, semiconductor grade MOS PURANAL (Honeywell 17938), ≥85%
H3O4P
40266
Phosphoric acid, semiconductor grade PURANAL (Honeywell 17861), ≥85%
H3O4P
612162
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
H3O4P
699608
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
H3O4P
698822
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
H3O4P
W290017
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
H3O4P
609781
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
H316O4P
RDD007
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
H2NaO4P
S8282
Sodium phosphate monobasic, BioXtra, ≥99.0%
H2NaO4P
S3139
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
H2NaO4P
S5011
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
H2NaO4P
S2554
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
H2NaO4P
S6566
Sodium phosphate monobasic, Biotechnology Performance Certified, Cell Culture Tested
H2NaO4P
229903
Sodium phosphate monobasic, 99.999% trace metals basis
H2NaO4P
S0751
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
H2NaO4P
74092
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
H2NaO4P
609773
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
H2Na16O4P