EMAIL THIS PAGE TO A FRIEND
Eukaryotic Cell
Interplay between Gliotoxin Resistance, Secretion, and the Methyl/Methionine Cycle in Aspergillus fumigatus.
PMID 26150413
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
463035
Methane, electronic grade, ≥99.998%
CH4
295477
Methane, ≥99.0%
CH4