EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Structure-Based Design of Macrocyclic Coagulation Factor VIIa Inhibitors.
PMID 26151189
Abstract
On the basis of a crystal structure of a phenylpyrrolidine lead and subsequent molecular modeling results, we designed and synthesized a novel series of macrocyclic FVIIa inhibitors. The optimal 16-membered macrocycle was 60-fold more potent than an acyclic analog. Further potency optimization by incorporation of P1' alkyl sulfone and P2 methyl groups provided a macrocycle with TF/FVIIa Ki = 1.6 nM, excellent selectivity against a panel of seven serine proteases, and FVII-deficient prothrombin time EC2x = 1.2 μM. Discovery of this potent, selective macrocyclic scaffold opens new possibilities for the development of orally bioavailable FVIIa inhibitors.