EMAIL THIS PAGE TO A FRIEND
Mycoses
Evaluation of virulence factors and antifungal susceptibility patterns of different Candida species isolated from the female camel (Camelus dromedarius) genital tract.
PMID 26152466
Abstract
The purposes of this study were to investigate the enzymatic activity of different Candida species and their antifungal susceptibility patterns. The study involved a total of 83 isolates of Candida from the genital tract of the female Camelus dromedarius. After species identification, the isolates were analysed for the production/activity of phospholipase, proteinase and haemolysin. In addition, the agar disc diffusion method was performed on the basis of CLSI guidelines M44-A2 protocol for antifungal susceptibility testing. All the isolates were able to produce phospholipase, proteinase and haemolysin. A total of 35.48%, 87.09% and 64.51% of C. albicans isolates exhibited very high phospholipase, proteinase and haemolytic activities, respectively, whereas very high phospholipase, proteinase and haemolytic activities were determined in 5.76%, 23.07% and 45.16% of non-C. albicans isolates respectively. Overall, 61 (73.5%) of Candida isolates were susceptible to fluconazole, 70 (84.3%) susceptible to clotrimazole, 82 (98.8%) susceptible to voriconazole, 76 (91.6%) susceptible to itraconazole, 75 (90.4%) susceptible to ketoconazole, 83 (100%) susceptible to amphotericin B, 81 (97.6%) susceptible to nystatin and 36 (43.4%) susceptible to flucytosine. Candida isolates showed higher haemolytic activity than that of other secreted hydrolases among vaginal Candida species. In addition, amphotericin B was the most in vitro effective antifungal drug and flucytosine had the poorest activity under such conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7129
5-Fluorocytosine, nucleoside analog
C4H4FN3O