EMAIL THIS PAGE TO A FRIEND
Respiratory Care
Pressurized Metered-Dose Inhalers Versus Nebulizers in the Treatment of Mechanically Ventilated Subjects With Artificial Airways: An In Vitro Study.
PMID 26152470