EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Catalytic Mechanisms of Fe(II)- and 2-Oxoglutarate-dependent Oxygenases.
PMID 26152721
Abstract
Mononuclear non-heme Fe(II)- and 2-oxoglutarate (2OG)-dependent oxygenases comprise a large family of enzymes that utilize an Fe(IV)-oxo intermediate to initiate diverse oxidative transformations with important biological roles. Here, four of the major types of Fe(II)/2OG-dependent reactions are detailed: hydroxylation, halogenation, ring formation, and desaturation. In addition, an atypical epimerization reaction is described. Studies identifying several key intermediates in catalysis are concisely summarized, and the proposed mechanisms are explained. In addition, a variety of other transformations catalyzed by selected family members are briefly described to further highlight the chemical versatility of these enzymes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
378704
Tropane, 98%
C8H15N