EMAIL THIS PAGE TO A FRIEND
Tissue Engineering: Part C, Methods
Embedded 3D Photopatterning of Hydrogels with Diverse and Complex Architectures for Tissue Engineering and Disease Models.
PMID 26154197