EMAIL THIS PAGE TO A FRIEND
International journal of environmental research and public health
Effects of Acute Ingestion of Native Banana Starch on Glycemic Response Evaluated by Continuous Glucose Monitoring in Obese and Lean Subjects.
PMID 26154657
Abstract
An abnormal glycemic profile, including postprandial glycemia and acute glucose spikes, precedes the onset of overt diabetes in obese subjects. Previous studies have shown the beneficial effects of chronic native banana starch (NBS) supplementation. In this study, we examined the effects of acute ingestion of NBS on glycemic profiles by means of continuous glucose monitoring in obese and lean subjects. In a crossover study, obese and lean subjects consumed beverages containing either 38.3 g of NBS or 38.3 g of digestible corn starch (DCS) twice daily during 4 days. On day 5, a 3-h meal tolerance test (MTT) was performed to evaluate glucose and insulin responses. After 1 week of washout period, treatments were inverted. NBS supplementation reduced the 48-h glycemia AUC in lean, obese, and in the combined group of lean and obese subjects in comparison with DCS. Postprandial glucose and insulin responses at MTT were reduced after NBS in comparison with DCS in all groups. However, no changes were observed in glycemic variability (GV) indexes between groups. In conclusion, acute NBS supplementation improved postprandial glucose and insulin responses in obese and lean subjects during 48 h of everyday life and at MTT. Further research to elucidate the mechanism behind these changes is required.