Indian journal of experimental biology
Down-regulation of osmotin (PR5) gene by virus-induced gene silencing (VIGS) leads to susceptibility of resistant Piper colubrinum Link. to the oomycete pathogen Phytophthora capsici Leonian.
PMID 26155671
Piper colubrinum Link., a distant relative of Piper nigrum L., is immune to the oomycete pathogen Phytophthora capsici Leonian that causes 'quick wilt' in cultivated black pepper (P. nigrum). The osmotin, PR5 gene homologue, earlier identified from P. colubrinum, showed significant overexpression in response to pathogen and defense signalling molecules. The present study focuses on the functional validation of P. colubrinum osmotin (PcOSM) by virus induced gene silencing (VIGS) using Tobacco Rattle Virus (TRV)-based vector. P. colubrinum plants maintained under controlled growth conditions in a growth chamber were infiltrated with Agrobacterium carrying TRV empty vector (control) and TRV vector carrying PcOSM. Three weeks post infiltration, viral movement was confirmed in newly emerged leaves of infiltrated plants by RT-PCR using TRV RNA1 and TRV RNA2 primers. Semi-quantitative RT-PCR confirmed significant down-regulation of PcOSM gene in TRV-PcOSM infiltrated plant compared with the control plants. The control and silenced plants were challenged with Phytophthora capsici which demonstrated that knock-down of PcOSM in P. colubrinum leads to increased fungal mycelial growth in silenced plants compared to control plants, which was accompanied by decreased accumulation of H2O2 as indicated by 3,3'-diaminobenzidine (DAB) staining. Thus, in this study, we demonstrated that Piper colubrinum osmotin gene is required for resisting P. capsici infection and has possible role in hypersensitive cell death response and oxidative burst signaling during infection.
Related Materials
Product #
Molecular Formula
Add to Cart
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%