EMAIL THIS PAGE TO A FRIEND
Tropical animal health and production
Molecular characterization of the circulating Bacillus anthracis in Jordan.
PMID 26156620
Abstract
To understand the biomolecular charcteristics of Bacillus anthracis in Jordan, 20 blood smear slides from dead animals with suspected anthrax were analyzed using conventional and molecular approaches. All slides were positive for B. anthracis by conventional staining but no growth of the organism on selective media was detected. However, of the 20 samples, 16 were B. anthracis DNA-positive using polymerase chain reaction (PCR). Seven samples provided enough quantity and quality of DNA, and their multilocus variable tandem repeat analysis (MLVA)-15 loci analysis revealed two different genotypes. All genotypes were belonging to A.B..r. 008/009 which is very common in Asia and Europe. Single nucleotide repeat (SNR) analysis revealed that there were no sub genotypes. Molecular diagnosis of animal anthrax in Jordan is not used routinely; henceforth, official diagnosis of anthrax is based on the observation of the slides by optical microscope and this can often cause reading errors. Therefore, the prevalence of the disease in Jordan might be slightly lower than that reported by the official bodies.