EMAIL THIS PAGE TO A FRIEND
Journal of Clinical Microbiology
Infectious Middle East Respiratory Syndrome Coronavirus Excretion and Serotype Variability Based on Live Virus Isolates from Patients in Saudi Arabia.
PMID 26157150